Learn More
Witeg™ Micropipettes
Click to view available options
Volume (Metric):
20 to 200 μL
200 to 1000 μL
Unit Size:
Each
Description
- High quality pipette, made in Germany
- Economically priced
- Perfect for precise laboratory applications
- 7 color-coded single-channel models
- High accuracy and precision according to DIN EN ISO 8655-2
- Ergonomically designed and low weighted
- Optimized spring force for comfortable pipetting
- Robust and stable construction
- Tip ejector and piston made of stainless steel
- Suitable for all established brands of pipette tips
- Completely autoclavable at 121°C (5000mL filter is not autoclavable)
- Individual certificate and serial number
- DE-M marked according to German calibration law
Specifications
Specifications
| Accuracy | 1.6μL |
| ClipTip | Ejector Tip |
| Autoclavable | Autoclavable |
| Color | Yellow |
| For Use With | Precision Dispensing |
| Number of Channels | 1 |
| Type | Micropipette |
| Volume (Metric) | 20 to 200 μL |
| For Use With (Application) | Precision dispensing |
| Precision | ≤ 0.6 μL |
| Show More |