Learn More
Thermo Scientific™ Microplate Rack Inserts for MaxQ™ 8000 Shakers
Shop All Thermo Scientific ProductsDescription
Maximize efficiency in the lab by using Thermo Scientific™ Microplate Rack Inserts for MaxQ™ 8000 Shakers, which allow the shaking of multiple microwell plates at one time. With ultimate flexibility, these microplate rack inserts are the optimal choice for high throughput labs.
Specifications
Specifications
| Holds | Deep-well Plates |
| Product Type | Microplate rack inserts, deep-well plates |
| Description | Microplate rack inserts, deep-well plates |
| Packaging Quantity | 5 Racks, 30 plates |