Learn More
Ismatec™ Micropump™ Gear Pump Head 185
Click to view available options
Flow Rate:
1 to 100 mL/min
5 to 504 mL/min
Unit Size:
Each
Description
- Suction shoe design provides higher delivery pressures and temperatures, plus increased efficiency.
- Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery.
- The magnetic drive and static seals keep fluids securely inside and potential contaminants out.
- Chemical resistant materials handle a wide range of materials (due to wetted parts).
- Pump heads are compact and easy to service.
- The modular design offers flexibility—configure to your specific needs.
Specifications
Specifications
| Flow Rate | 5 to 504 mL/min |