Learn More
Sartorius Microsart™ E.jet Quick Connection Set, POM, PSU
Product Code. 15585253 Shop All Sartorius Products
Change view
Click to view available options
:
Microsart™ E.jet Quick Connection Set, POM, PSU
Product Code.
15585253 Microsart™ E.jet Quick Connection Set, POM, PSU
1 options
Product Code. 15585253
Brand: Sartorius 1EAS0027
Only null left
Add to basket
Add to basket
Microsart™ E.jet Quick Connection Set - streamline laboratory productivity.
  • 2 Nipples (POM) on R3/8 in. male thread and 2 Couplings (PSU) on DN 10 hose nipple
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
For Use With (Equipment) Microsart e.jet Transfer Pump
Sterility Non-sterile
Type Connector