Learn More
Thermo Scientific™ Microscope Slides, Cut, Square
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
Brand:  Thermo Scientific™ AAAC0220076002EB0MNZ#0#G#
Additional Details : Poids : 0.45000kg
 View more versions of this product
Product Code. 15642239
Cet article n'est plus commercialisé et n'est donc pas disponible. Consultez le produit pour voir les alternatives possibles ou contactez notre support technique au 056 260 260.
Description
Description
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications
Specifications
76mm
1mm
Extra White
100 pieces in paper
76 x 22mm
Soda-lime glass
22mm
Uncoated
No