Learn More
Greiner Bio-One Microscopic Cover slip for Terasaki Plates
Product Code. 11719502
Click to view available options
:
Microscopic coverslip
Product Code. 11719502
Brand: Greiner Bio-One 653081
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For use with Terasaki plates. Greiner Bio-One Microscopic Cover slip is manufactured from crystal clear polystyrene.
  • Manufactured from crystal clear polystyrene
  • Non-sterile
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Length (Metric) 76 mm
Width (Metric) 51 mm
For Use With (Equipment) Terasaki Plates