Learn More
IBA Lifesciences Strep-Tactin™ Coated Microplate
Shop All IBA Lifesciences Products
Click to view available options
Surface Treatment:
Strep-Tactin
Unit Size:
Each
Description
- Ready-to-use Strep-Tactin™-coated microplate (12 x 8-well strips) for ligand binding assays, immunoassays, enzyme assays, or diagnostic assays under high throughput conditions.
- Due to the high specificity and affinity (μM-nM) of the streptavidin variant Strep-Tactin™, Strep-tag™II or Twin-Strep-tag™ fusion proteins can be selectively and stably captured even from complex mixtures.
- Biomolecules of interest are presented to interaction partners in a uniform manner, resulting in reliable and reproducible assays.
- The solid-phase, multi-well format is compatible with standard multichannel pipettes and automated plate readers.
- Twin-Strep-tag™ fusion protein binding capacity determined with bacterial alkaline phosphatase (97 kDa) is 2 pmol for 200 μl incubation volume.
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| No. of Wells | 96 |
| Surface Treatment | Strep-Tactin |
| Total Volume (Metric) Well | 300 μL |
| Quantity | 1 Pk. |
| Temperature Range (Metric) | 4°C |