Learn More
Corning™ Microvolume Pipet Tips, Nonsterile
Product Code. 10479131 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.2 to 10 μL
0.5 to 10 μL
Format:
Bulk
Rack
Unit Size:
Pack of 10000
Pack of 960
Product Code. 10479131
Brand: Corning™ 4834
Only null left
Add to basket
Add to basket
Provide accurate, reliable performance in the stated volume range
  • Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
  • RNase-/DNase-free.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.5 to 10 μL
Sterility Non-sterile
Color Natural
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Microvolume, Round
Packaging Rack
For Use With (Equipment) Eppendorf, Universal Fit Ultramicro
Material Polypropylene