missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Mini Sampler Case

Product Code. 15829521 Shop All Sampling Systems Products
Click to view available options
Dimensions (L x W x H):
250 x 175 x 40 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15829521

Brand: Sampling Systems™ 8505A12

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Sampling Systems range of heavy duty cases are ideal for storing samplers.

  • Ideal for storing small samplers
  • Simple clasps keep the case closed
  • Economic
  • Supplied with a sturdy carrying handle

Specifications

Product Type Storage Case
Color Black
For Use With (Application) Storing samplers
Dimensions (L x W x H) 250 x 175 x 40 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.