Description
- Pump only
- For use with Buerkle™ MiniSampler
Specifications
Specifications
| Product Type | Vacuum Pump |
| Material | Polypropylene |
| Color | Black and Yellow |
| For Use With (Equipment) | Buerkle MiniSampler |