Learn More
Edmund Buhler Battery Shaker KM 2 Akku
Product Code. 10439971
Change view
Click to view available options
Speed Range:
30 to 420 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range unitSize
10439971 30 to 420 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10439971 Supplier Edmund Buhler Supplier No. 6135 000
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Rich, robust, reliable Battery Shaker 2 Akku that is ideal for various tasks due to its off-grid battery with a run time of 40 hours.
  • The Battery Shaker KM 2 Akku with orbital motion can be used for various tasks due to its off-grid battery with a run time of min
  • 40 hours
  • The shaking platform of 200 x 295 mm can be loaded with maximum 2 kg
  • The stroke of the laboratory shaker KM 2 Akku is 20 mm and the shaking speed is between 5 and 220 rpm
Specifications
Orbit 8 mm
Load Bearing Capacity (Metric) 2 kg
Speed Range 30 to 420 rpm
Electrical Requirements 100/240 V, 50/60 Hz
Temperature Range (Metric) 5°C to 40°C
Holds 20 x 10 mL Erlenmeyer Flasks/12 x 50 mL Erlenmeyer Flasks
Product Type Miniature Shaker
Frequency 50/60 Hz
Depth (Metric) 410 mm
Height (Metric) 130 mm
Width (Metric) 220 mm
Weight (Metric) 9.5 kg
Model KM 2 Akku
Display Digital
For Use With (Equipment) For operation in refrigerators/incubators
Humidity Range ∼85%
Includes Basic platform and Rubber mat, with battery charger/power unit
Timer Range Continuous operation, additional timer on sector model 120 min.