missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva MiniBeads™ Columns
Shop All Cytiva Products
Click to view available options
Product Type:
Mini Q Ion Exchange Column
Mini S Ion Exchange Column
Bed Volume:
0.24 mL
0.8 mL
Unit Size:
Each
Description
- Small, mono-disperse beads for extremely high resolution purifications. non porous beads contribute to high resolution but give lower capacity than MonoBeads
- Analytical and micro-preparative applications with choice of both anion and cation exchangers
- High chemical and physical stability
- Excellent reproducibility and durability
- Tricorn high-performance columns (4.6/50 PE) prepacked with MiniBeads
- PC columns designed for micropurification
Precision column holders may be required for use on high performance chromatography systems such as KTA design. For optimum results, it is important to check the compatibility of the column and the system specifications.
Specifications
Specifications
| Buffer | All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid |
| Product Type | Mini S Ion Exchange Column |
| Bed Volume | 0.24 mL |
| Diameter (English) Inner | 0.12 in. |
| Diameter (Metric) Inner | 3.2 mm |
| Quantity | Each |
| Height (English) | 1.96 in. |
| Height (Metric) | 50 mm |
| Flow Rate | 0.1 to 1 mL/min. |
| Includes | MiniBeads |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction