missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MiniBeads™ Columns

Product Code. 10257754 Shop All Cytiva Products
Click to view available options
Product Type:
Mini Q Ion Exchange Column
Mini S Ion Exchange Column
Bed Volume:
0.24 mL
0.8 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10257754

Brand: Cytiva 17068601

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

MiniBeads™ prepacked columns are designed for high performance purification and characterization of biomolecules using ion exchange chromatography

  • Small, mono-disperse beads for extremely high resolution purifications. non porous beads contribute to high resolution but give lower capacity than MonoBeads
  • Analytical and micro-preparative applications with choice of both anion and cation exchangers
  • High chemical and physical stability
  • Excellent reproducibility and durability

  • Tricorn high-performance columns (4.6/50 PE) prepacked with MiniBeads
  • PC columns designed for micropurification

Precision column holders may be required for use on high performance chromatography systems such as KTA design. For optimum results, it is important to check the compatibility of the column and the system specifications.

TRUSTED_SUSTAINABILITY

Specifications

Buffer All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid
Product Type Mini Q Ion Exchange Column
Bed Volume 0.24 mL
Diameter (English) Inner 0.12 in.
Diameter (Metric) Inner 3.2 mm
Quantity Each
Height (English) 1.96 in.
Height (Metric) 50 mm
Flow Rate 0.1 to 1 mL/min.
Includes MiniBeads
Material Glass
Ligand Type O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3
Max. Pressure 100psi
pH Range 3 to 11
Particle Size 3 μm
Support Type Monodisperse Polystyrene/Divinyl Benzene particles
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Microscale final polishing and analytical ion exchange separations
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.