Learn More
Lovibond™ Water Testing Minikit
Product Code. 10248520
Click to view available options
Range:
0 to 500 mg/L
0.75 to 10% acid
10 to 500 mg/L
2 to 50 mg/L
20 to 200 mg/L
20 to 500 mg/L
5 to 5000 mg/L
70 to 1500 mg/L
Analytes:
Alkalinity Total M
Alkalinity-P, CaC03
Chloride
Cleaning Acid Strength
Cyanuric Acid
Nitrite
Sulphate (low range) Na2SO4
Sulphite (high range)
Sulphite (low range)
Unit Size:
Each
Product Code. 10248520
Brand: Lovibond™ undefined
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
  • Easy operation and exact reagent dosing
  • High accuracy
  • Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
  • Unrestricted shipment
  • Safe storage
Specifications
Analytes Nitrite
Includes Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions.
Quantity 1 Pack
Range 70 to 1500 mg/L
Sufficient For Approximately 40 Tests
Test Method Tablet count
Type Minikit water testing kit