Learn More
Lovibond™ Minikit™ Sulphate
Brand:  Lovibond™ 414310
Additional Details : Weight : 0.21600kg
Product Code. 10472301
  • 77.70€ / Each
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
Specifications
For the Determination of Sulphate Levels between 40 and 200mg/L
1 Each
40 to 4000mg/L
Minikit Sulphate