Learn More
Cole-Parmer™ Digi-Sense Talking 1-Channel Digital Clock/Timer
Product Code. 12628103 Shop All Cole-Parmer Products
Click to view available options
Display:
LCD
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 12628103
Brand: Cole-Parmer™ 9446012
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under eco-contribution tax.
A clear human voice announces time remaining in count-down mode-no need to visually check
  • Announcements increase in frequency as time approaches zero
  • In clock mode, press any button and the voice will announce the time
  • Perfect for low-light applications, darkrooms, and activities that require your full attention
Specifications
Product Type LCD Timer
Alarms 1 minute (6 alarm sounds)
Display LCD
Battery Type 1.5 V
Width (Metric) 64 mm
Height (Metric) 83 mm
Quantity 1 Each