Learn More
Epredia™ Miscellaneous Microtome Section Waste Tray with Arm Rest, For HM 310, HM 315, HM325, and HM 335
Shop All Epredia Products
Click to view available options
Product Type:
Foot Pedal
Large-field Magnifier
Unit Size:
Each
Description
- Interlock plug replaces foot pedal connector
- Storage unit can be removed and used separately; tray for specimens can be cooled by changeable inserts
- Large-field magnifier attaches to facilitate knife alignment and approach for sectioning; magnification 2.5X
Specifications
Specifications
| Product Type | Large-field Magnifier |
| For Use With (Equipment) | Epredia 200/300 Series Microtomes |