Learn More
Emma Safety Footwear Mitchel Safety Shoes
Ideal for working with metal with optimal comfort, ventilation and moisture removal technology. A perfectly integrated steel toe protects feet against falling pieces of metal and other exterior threats. The Gripforce™ DUO soles withstand high temperatures and are oil and petrol resistant.
Brand:  Emma Safety Footwear MM15484844
Additional Details : Weight : 2.10000kg
 View more versions of this product
Product Code. 15938251
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction with extra wear-resistant coating
  • GripForce™ Duo Heat Resistant Outsole
  • Heat and cold insulated
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Black
Hydro-Tec™ Sanitized Silver
44
Laces
Mitchel
Leather
Steel