Learn More
Emma Safety Footwear Mitchel Safety Shoes
Ideal for working with metal with optimal comfort, ventilation and moisture removal technology. A perfectly integrated steel toe protects feet against falling pieces of metal and other exterior threats. The Gripforce™ DUO soles withstand high temperatures and are oil and petrol resistant.
Brand: Emma Safety Footwear MM15484845
Additional Details : Weight : 2.15300kg
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction with extra wear-resistant coating
- GripForce™ Duo Heat Resistant Outsole
- Heat and cold insulated
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
45 |
Laces | |
Mitchel | |
Leather | |
Steel |