Learn More
Esculape Laboratoire™ Mix Eyestation
Use Esculape Laboratoire™ Mix Eyestation within six months of its placement since it expires and requires a bi-annual maintenance schedule. These units must be cleaned and refilled in accordance with the manufacturer’s instructions.
Brand:  Esculape Laboratoire™ 8205205
Additional Details : Weight : 0.80000kg
Product Code. 11854021
  • 146.28€ / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • Mixed preservative with tap water
  • ontinuous flushing with 0.4 gallons per minute for 15 minutes
  • Hands-free operation allows user to hold both eyes open simultaneously
Specifications
Specifications
Washing the Eyes in the Event of Splashes of Acids or Bases
Eye wash bottle with 200 mL neutral pH solution; Eye wash bottle with 500 mL of ophthalmic solution; Box for wall mounting with mirror and user manual
7 min.