Learn More
Merck Mixed Cellulose Ester Gridded Surface Membrane Filters
Brand: Merck HAWG05000
Additional Details : Weight : 0.10000kg
Specifications
White | |
MCE | |
For laboratory filtration applications | |
Gridded | |
50 mm |
Membrane Filter | |
0.45 μm | |
100 Pack | |
Membrane Filter |