missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 154
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 154 is a potent neuropeptide S receptor (NPSR) antagonist (pA2 = 9.98). Inhibits neuropeptide S-induced ERK phosphorylation over cAMP responses and calcium responses (IC50 values are 9.3, 22.1 and 36.5 nM, respectively). Appears to modulate addictive behavior in vivo. Displays no activity against vasopressin V1B receptors. Brain penetrant.
Specifications
Specifications
| Purity | >99% |
| Quantity | 10 mg |
| Product Type | ML 154 |
| Molecular Weight (g/mol) | 545.47 |
| Inhibitors | ML 154 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction