missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 184

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 184 is a selective GPR55 agonist (EC50 = 250 nM). Exhibits >100-fold selectivity for GPR55 over GPR35, CB1 and CB2. Increases proliferation of neural stem cells and promotes neuronal differentiation in vitro.
Specifications
Specifications
| Synonym | CID 20440433 |
| Quantity | 10 mg |
| Molecular Formula | C25H34N4O3S |
| Formula Weight | 470.63 |
| Product Type | Agonist |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction