missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 351
GREENER_CHOICE

Product Code. 18793198
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18793198

Brand: Tocris Bioscience™ 6448/5

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Selective 12/15 LOX inhibitor; active in vivo

ML 351 is a selective 12/15 LOX inhibitor (IC50 = 200 nM). Exhibits >250-fold selectivity over related isozymes, 5-LOX, platelet 12-LOX, 15-LOX-2, ovine COX-1, and human COX-2. Protects against oxidative glutamate toxicity in mouse neuronal cells (HT-22) and reduces infarct size in a mouse ischemic stroke model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(Methylamino)-2-(1-naphthalenyl)-4-oxazolecarbonitrile
CAS 847163-28-4
Quantity 5 mg
Molecular Formula C15H11N3O
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.