missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 354
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 354 is a selective PAR4 antagonist (IC50 = 140 nM). Exhibits 70-fold selectivity for PAR4 over PAR1.
Specifications
Specifications
| Purity | >98% |
| Description | ML 354 |
| Molecular Weight (g/mol) | 282.29 |
| Product Type | ML 354 |
| Quantity | 10 mg |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction