Learn More
Tocris Bioscience™ ML 365
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 365 is a potent and selective TASK-1 (K2P3.1/KCNK3) channel blocker (IC50 values are 4 and 390 nM at TASK-1 and TASK-3, respectively). Displays little or no inhibition at Kir2.1, voltage-gated potassium channels, KCNQ2 and KV11.1 (hERG).
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | ML 365 |
| Purity | >99% |
| Molecular Weight (g/mol) | 360.41 |
| Inhibitors | ML 365 |