missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 385

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
ML 385 is an Nrf2 inhibitor. Blocks Nrf2 downstream target gene expression. Enhances cytotoxic effects of doxorubicin (Cat. No. 2252) or Taxol (Cat. No. 1097) in non-small cell lung cancer (NSCLC). Demonstrates selectivity for NSCLC cells with KEAP1 mutations.
Specifications
Specifications
| Quantity | 5 mg |
| Molecular Formula | C29H25N3O4S |
| Formula Weight | 511.59 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction