missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MMP1-Cell Lysate ELISA Kit

Product Code. 17707983
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17707983 10 x 96 Tests Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17707983 Supplier Invitrogen™ Supplier No. EHMMP1CLX10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P03956
Assay Range 24.69 to 18,000 pg/mL
Assay Sensitivity 8 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4312
Gene Symbol MMP1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Cell Lysis Buffer, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 20 μL
Storage Requirements 2°C to 8°C
Target MMP1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.