Learn More
IKA MMT 100.1, Multi-use Milling Tube 100ml
Brand: IKA 0020016315
290.94 EUR valid until 2026-05-31
BEST PRICE on Promo! Use promo code "27998" to get your promotional price.
MMT 100.1, Multi-use Milling Tube 100ml
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.