Learn More
SGE™ MN Silicone Septa
Click to view available options
Diameter (Metric) Outer:
11 mm
Unit Size:
Pack of 50
Description
- MN silicone Premium septum
- For autosamplers with up to 400 injections per septum
Specifications
Specifications
| Type | Septum |
| Diameter (Metric) Outer | 11 mm |
| Material | Silicone |
| Quantity | 50 Pack |