Learn More
IKA™ R 2850 Mobile Floor Stand
Product Code. 15251136 Shop All IKA Products
Click to view available options
Diameter (Metric):
16 mm
Unit Size:
Each
Product Code. 15251136
Brand: IKA™ 0020002900
Only null left
Add to basket
Add to basket
Mobile floor stand, with H-shape base which prevents against tipping.
For overhead stirrers and dispersers with a diameter of extension arm of 16 mm.
TRUSTED_SUSTAINABILITY
Specifications
Depth (Metric) 875 mm
Diameter (English) 0.6 in.
Diameter (Metric) 16 mm
For Use With (Equipment) For Overhead Stirrers and Dispersers With a Diameter of Extension Arm of 16mm
For Use With (Application) For Overhead Stirrers and Dispersers With a Diameter of Extension Arm of 16mm
Height (Metric) 1921 mm
Quantity 1 Ea.
Type Mobile Floor Stand
Width (English) 38.6 in.
Width (Metric) 980 mm