Learn More
Emma Safety Footwear Modena Safety Shoes
Water resistant leather uppers, zero penetration in-sole and a steel safety toe cap. Offering marginally more ankle support, the Modena is a shoe that's specifically designed for standing, walking and the occasional run.
Brand: Emma Safety Footwear MM13609039
Additional Details : Weight : 1.57700kg
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- Nitrile Rubber Heat Resistant Outsole
- ESD Certified
- Chrome-free lining
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver & Wellness leather | |
39 |
Laces | |
Modena | |
Leather | |
Steel |