missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Block

Material: Anodized Aluminum; For Use With: Dry block heater

Brand:  OHAUS™ 30400185

Additional Details : Weight : 1.00000kg

Product Code. 15772599

  • 134.00€ / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket

Alert:

To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.


Description

Description

Modular blocks are sold separately from heaters.

  • Close contact of tubes-to-block-walls allow for maximum heat retention
  • Each block has a thermometer well for measuring block temperature

Specifications

Specifications

Module Block For Vials 19 mm
Anodized Aluminum
Block
1 Block
9.5 cm
5.1 cm
4.5 cm
1.97 cm
Dry block heater
12
19mm Vial
3.75 in.
2 in.
1.77 in.
0.77 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions