Learn More
Miele Professional™ Mobile Injector, Module A 300
Module A 300 for the optimum loading of laboratory glassware
Brand: Miele Professional™ 11116900
Additional Details : Weight : 2.00700kg
Description
- Suitable for laboratory glassware such as Erlenmeyer flasks
- equipped with 8 injector nozzles E 352 (6 x 220 mm)
- equipped with 8 locking mechanisms for nozzles E 354
- Jet spacing 120 mm across the width, 123 mm across the depth
Specifications
Injector Module | |
Laboratory washer baskets A 100 and A 150 | |
Two rows of four injectors | |
181 x 220 x 479mm |