Learn More
Thermo Scientific™ Molecular BioProducts™ PCR Tubes and Plates
Molded from 100% virgin polypropylene
POA
Description
Molded from 100% virgin polypropylene
Tubes feature thin walls for optimal heat transfer and caps that seal securely to minimize evaporation. Plates feature elevated well rims to ensure uniform sealing.
- PurePak™ packaging reduces contamination from reaching into a bulk bag of tubes by dividing tubes into 10 separate PurePaks
- Certified RNase-, DNase-, DNA, and Pyrogen-free
- Clear, nonsterile
Specifications
Thin Wall PCR Tube | |
Polypropylene |