missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 488 Conjugate

Product Code. 11504896
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Product Code. Quantity unitSize
11504896 500 μg 500µg
1 options
This item is not returnable. View return policy

Product Code. 11504896

Brand: Invitrogen™ I21411

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolated from the seeds of Griffonia simplicifolia, a tropical African legume

Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B, IA2B2, IAB3, and IB4). The A subunit prefers N-acetyl-D-galactosamine end groups while the B subunit is selective for terminal a-D-galactosyl residues. The green fluorescent Alexa Fluor 488 isolectin GS-IB4 conjugate can be used in fluorescence microscopy or flow cytometry.

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbial Tracking

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 μg
Source Africa Shrub legume
Conjugate Alexa Fluor 488
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Lectins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.