Learn More
Invitrogen™ BrUPT (5-Bromouridine 5´-Triphosphate), 10 mM in TE buffer
Monitor nucleolar transcription in situ
Brand: Invitrogen™ B21551
Additional Details : Weight : 0.00450kg
Description
BrUTP is an excellent substrate for RNA polymerase and has been used to monitor nucleolar transcription in situ. BrUTP can be detected with anti-BrdU antibodies.
Cell Analysis, Cell Proliferation, Cell Viability, Proliferation and Function
Order Info
Shipping Condition: Wet ice
Specifications
BrUPT | |
Wet Ice | |
BrdU |
Store in freezer -5°C to -30°C and protect from light. | |
10 mM | |
25 μL |
For Research Use Only. Not for use in diagnostic procedures.