Learn More
Invitrogen™ Calcium Calibration Buffer Kit #1, zero and 10 mM CaEGTA
Determine the dissociation constant (Kd) of fluorescent Ca2+ indicators using a fluorometer or quantitative imaging system
Brand: Invitrogen™ C3008MP
Additional Details : Weight : 0.14000kg
Description
- Kd of indicator can be calculated from plot generated by scanning excitation or emission of indicator in presence of eleven different Ca2+ concentrations
- Calcium Calibration Kit #1 contains 50mL of 10mm K2EGTA and 50mL of 10mm CaEGTA
- Both solutions contain 100mm KCl and 30mm MOPS, pH 7.2 and are prepared in deionized water
- These stock solutions can be blended to make buffers having free Ca2+ ranging from 0μM to 39μM
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Store in refrigerator at 2°C to 8°C | |
Calcium Calibration | |
Calcium Calibration Buffer Kit | |
Tube(s), Slide(s) |
Fluorescent | |
Fluorescence Microscope, Fluorometer | |
Other Label(s) or Dye(s) | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.