Learn More
Invitrogen™ Carboxy-PTIO (spin trap)
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
- Water-soluble carboxy-PTIO has been used as nitric oxide antagonist as well as to quantitate NO levels by ESR spectrometry
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitric Oxide Research, Nitro-Oxidative Stress
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 25 mg |
For Research Use Only. Not for use in diagnostic procedures.