Learn More
Invitrogen™ Click-IT™ Geranylgeranyl Alcohol, Azide, mixed isomers

Description
Identify and characterize geranylated proteins with Click-iT geranylgeranyl alcohol, azide using the powerful click chemistry, a simple and robust two-step labeling and detection technique. In step one, the azide-containing biomolecule is fed to cells or animals and actively incorporated into proteins. Unlike other labels such as biotin or a fluorescent dye, the azide-tag is small enough that the tagged molecule is an acceptable substrate for the enzymes that incorporate this building block into proteins. Detection utilizes the chemoselective ligation or “click” reaction between and azide and an alkyne where the modified protein is detected with the corresponding alkyne-containing dye or hapten using either the Click-iT Cell Reaction Buffer Kit or the Click-iT Protein Buffer Kit. With the Click-iT Cell Reaction Buffer Kit, cells can be analyzed by fluorescence microscopy, flow cytometry or high-content imaging and analysis (HCS) together with other biomarkers of interest for content and context rich results. With the Click-iT Protein Reaction Buffer Kit, achieve detection sensitivity in 1-D gels and western blots in the low femtomole range or perform LC-MS/MS and MALDI MS analysis.
- Less hazardous
Cell Analysis, Click Chemistries, Labeling Chemistry, Metabolic Labeling and Nascent Protein Synthesis with Click Chemistry
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Green Features | Less hazardous |
| Product Type | Geranylgeranyl Alcohol Azide |
| Quantity | 1 mg |
| Content And Storage | Store at ≤-20°C, desiccated and protected from light. |
| Labeling Method | Metabolic Labeling |
| Shipping Condition | Dry Ice |
| Product Line | Click-iT, Molecular Probes |
| Label or Dye | Azide |
| Format | Solid |
| Labeling Target | Proteins |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.