missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELF™ 97 Endogenous Phosphatase Detection Kit

Product Code. 10246332
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10246332 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10246332 Supplier Invitrogen™ Supplier No. E6601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be used to detect phosphatase activity

Upon hydrolysis the ELF 97 phosphatase substrate in the ELF 97 Endogenous Phosphatase Kit produces a bright and photostable yellow-green fluorescent precipitate at the site of enzymatic activity. This fluorescent precipitate has several unique spectral characteristics, including an extremely large Stokes shift, 180nm that makes it easily distinguishable from endogenous fluorescence.

Cell Analysis, Cellular Imaging, Drug Discovery & Development, Enzyme & Protein Activity Assays, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Immunohistochemistry (IHC), Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation & Analysis, Target & Lead Identification & Validation

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow-Green
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Phosphatase Assay
For Use With (Equipment) Fluorescence Microscope
Product Type Phosphatase Assay
Dye Type ELF 97
Emission 345/530
Form Liquid
Product Line ELF
Quantity 1 kit
Shipping Condition Wet Ice
Substrate ELF™ 97 phosphate
Substrate Properties Chemical Substrate
Substrate Type Phosphatase Substrate
Target Enzyme Phosphatases
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.