missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluo-5N, Pentapotassium Salt, cell impermeant

Product Code. 11514786
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
This item is not returnable. View return policy

Product Code. 11514786

Brand: Invitrogen™ F14203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Analog of fluo-4 with lower Ca2+-binding affinity, making it suitable for detecting intracellular calcium levels in 1μM to 1mM range that would saturate response of fluo-3 and fluo-4

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+.

  • Cells may be physically loaded with cell-impermeant salt forms of these indicators using patch pipet or microinjection or Influx™ pinocytotic cell-loading reagent
  • Compatible with excitation at 488nm by argon-ion laser sources, making them useful for confocal microscopy, flow cytometry, and microplate screening applications
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
  • BAPTA-based indicators bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+
  • Perturbations to calcium measurements caused by presence of the ions can be controlled using heavy metal-selective chelator

  • Label (Ex/Em of Ca2+-bound form): Fluo-5N (494/516nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in buffer: approximately 90μM

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
Product Type Calcium Indicator
Dye Type Fluorescent Dye-Based
Quantity 500 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.