missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lectin PNA From Arachis hypogaea (peanut), Alexa Fluor™ 647 Conjugate

Product Code. 10308072
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 10308072

Brand: Invitrogen™ L32460

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Specific for terminal β-galactose

Lectin PNA will agglutinate human erythrocytes, but only after neuraminidase treatment.[ALTREF PN40265]

Cell Analysis, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Cellular Imaging, Endocytosis, Exocytosis & Phagocytosis, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, In Vivo Imaging, Microbial Tracking

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Source Peanut
Conjugate Alexa Fluor 647
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Lectins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.