missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RediPlate™ 96 EnzChek™ Serine/Threonine Phosphatase Assay Kit

Product Code. 10145142
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10145142

Brand: Invitrogen™ R33700

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Detects serine/threonine phosphatases and their corresponding modulators and inhibitors

The RediPlate 96 EnzChek Serine/Threonine Phosphatase Assay Kit provides a fast, simple and direct fluorescence-based assay for detecting serine/threonine phosphatases and their corresponding modulators and inhibitors. Unlike other microplate assays, this kit provides the necessary reagents predispensed into a 96-well microplate. Simply reconstitute the fluorogenic substrate in the assay wells with buffer, add the desired sample to the wells, incubate and then quantitate the fluorescence in any standard fluorescence-based microplate reader. Inhibitors are included in each assay well to ensure that the assay is selective for Ser/Thr PPases- other phosphatases, including tyrosine phosphatase do not significantly react with the substrate. Additional advantages of this assay include compatibility with non-ionic detergents and insensitivity to free phosphate.

Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Phosphatase Assay
For Use With (Equipment) Microplate Reader
Product Type Phosphatase Assay
Dye Type DiFMU
Emission 358/452
Product Line EnzChek, RediPlate
Quantity 1 x 96-well Plate
Shipping Condition Room Temperature
Substrate DiFMUP
Substrate Properties Chemical Substrate
Substrate Type Phosphatase Substrate
Target Enzyme Serine⁄Threonine Phosphatases
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.