Learn More
Invitrogen™ Molecular Probes™ Zymosan A BioParticles™ Opsonizing Reagent
Specifically designed to enhance uptake of zymosan A BioParticles conjugates
Brand:  Invitrogen™ Z2850
Additional Details : Weight : 0.00450kg
Product Code. 10013282
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Derived from highly purified rabbit polyclonal IgG antibodies
Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Flow Cytometry, Flow Cytometry Endocytosis, Exocytosis and Phagocytosis Assays, Flow Cytometry of Cellular Processes
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
1 unit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.