Learn More
Molecular Sieve, SI Analytics™
Supplied in a glass bottle
Brand:  SI Analytics 285226010
Additional Details : Weight : 0.20000kg
Product Code. 16316465
  • 60.85€ / Each
Estimated Shipment: 28-05-2024
to see stock.
Description
Description
  • Approx. 200 g in 250 mL glass bottle
Specifications
Specifications
200 g