missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 4A, Pellets, Dia.: 1.6mm, Honeywell Fluka™

Catalog Number p-7120597
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
70955-01-0
C20H25FN2O8
440.42
MFCD00131613
FJUZEZRZKVMAMD-UHFFFAOYNA-N
73906282
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
This item is not returnable. View return policy
70955-01-0
C20H25FN2O8
440.42
MFCD00131613
FJUZEZRZKVMAMD-UHFFFAOYNA-N
73906282
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Pellets, 1.6 mm diameter

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color White to Brown
Density 2g/cm3
Packaging Poly Bottle
Particle Size Dia.: 1.6mm
Quantity 500 g
Physical Form Pellets
Melting Point 400°C
Linear Formula Na12[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
IUPAC Name Molecular sieves
Chemical Name or Material Molecular sieves, 4Å
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'