missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 4A, Pellets, Dia.: 3.2mm, Honeywell™ Fluka™

Catalog Number p-7120598
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Drum with inliner
Poly Bottle
This item is not returnable. View return policy
70955-01-0
C20H25FN2O8
440.42
MFCD00131613
FJUZEZRZKVMAMD-UHFFFAOYNA-N
73906282
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
This item is not returnable. View return policy
70955-01-0
C20H25FN2O8
440.42
MFCD00131613
FJUZEZRZKVMAMD-UHFFFAOYNA-N
73906282
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Pellets, 3.2 mm diameter

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color Brown
Density 2g/cm3
Packaging Poly Bottle
Particle Size Dia.: 3.2mm
Quantity 1 kg
Physical Form Pellets
Melting Point 400°C
Linear Formula Na12[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
Chemical Name or Material Molecular sieves, 4Å
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'