missing translation for 'onlineSavingsMsg'
Learn More

Monkeypox Virus A35 Antibody (0026), Alexa Fluor™ 750 , Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-18812AF750

535.50 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18570559

  • 625.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Monkeypox Virus A35 Monoclonal antibody specifically detects Monkeypox Virus A35 in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Monkeypox Virus A35
Monoclonal
Alexa Fluor 750
Mouse
Protein A purified
RUO
Virus
Purified
Western Blot, ELISA
0026
50mM Sodium Borate
This product is a recombinant monoclonal antibody made to recombinant Monkeypox Virus A35 and expressed from HEK293 cells.
0.1 mL
Primary
Store at 4°C in the dark.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.