Learn More
Monomethylamine solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
BIOPHARMA
Product Code. p-7083620 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
500mL
Product Code. 12942155
Brand: Fisher Chemical A 029 100
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Chemical Identifiers
Molecular Formula CH3NH3+
Specifications
Packaging Bottle
Physical Form Liquid
Concentration 1 g/L
Type Single-component Standard Solution
Solvent or Matrix Water
Quantity 100 mL
Chemical Name or Material Monomethylamine